Zirconyl nitrate hydrate 99.5% contains up to 4% Hf

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate

AC22263-1000 AC22263-5000
TS22263-1000EA 104 USD
TS22263-1000 TS22263-5000
Zirconyl nitrate hydrate 99.5% contains up to 4% Hf
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
MW: 249.25 g/mol
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 3085
ADR: 5.1 III

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Degree of hydration 6 to 7
Gravimetry 34.9 to 37.0 % (ZrO2) (calculated on the actual degree of hydration)
Hafnium (Hf) ≤4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR