Zirconyl chloride octahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zirconium(IV) oxide chloride octahydrate, Zirconium dichloride oxide octahydrate, Zirconium(IV) oxychloride octahydrate
AC20837-0050
AC20837-1000
AC20837-5000
TS20837-0050EA
20
USD
TS20837-0050
TS20837-1000
TS20837-5000
Zirconyl chloride octahydrate 98%
Zirconyl chloride octahydrate
Formula:
Cl₂OZr·8H₂O MW: 322.25 g/mol Melting Pt: > 150 °C Density: 1.91 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149898 CAS Number: 13520-92-8 EINECS: 231-717-9 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Titration Argentometric | 97.5 to 102.5 % |
Water | 42 to 50 % |
Solubility | (10% in water) Clear to slightly hazy solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...