Sodium sulfide hydrate 60 - 64%, scales, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: di-Sodium sulphide hydrate

AC13372-0010 AC13372-0025 AC13372-0100
TS13372-0010EA 57 USD
TS13372-0010 TS13372-0025 TS13372-0100
Sodium sulfide hydrate 60 - 64%, scales, Extra Pure
Sodium sulphide hydrate
Formula: Na₂S·xH₂O
MW: 96.06 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00149183
CAS Number: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8,II
REACH: 01-2119513694-38
Merck Index: 12,08829

Order Now

Specification Test Results

Appearance Yellow Flakes
Titration Iodimetric 60 to 64 % (Na2S)
Sodium carbonate (Na2CO3) ≤1.2 % (Na2CO3)
Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR