Tripotassium orthophosphate monohydrate 96%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium phosphate monohydrate, Potassium phosphate tribasic monohydrate, Potassium phosphate hydrate

AC26379-0010 AC26379-0050 AC26379-0250 AC26379-0251 AC26379-5000
TS26379-0010EA 95 USD
TS26379-0010 TS26379-0050 TS26379-0250 TS26379-0251 TS26379-5000
Tripotassium orthophosphate monohydrate 96%, Extra Pure
Tripotassium orthophosphate monohydrate
Formula: K₃O₄P·1H₂O
MW: 230.28 g/mol
Melting Pt: 1340 °C
Storage Temperature: Ambient
MDL Number: MFCD00150410
CAS Number: 27176-10-9
EINECS: 231-907-1

Order Now

Specification Test Results

Appearance White Powder
Titration with HClO4 ≥95 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm
Loss on ignition ≤13 % (800°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR