Hexachloroplatinic(IV) acid hydrate (38 - 40% Pt) 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate
AC36359-0010
AC36359-0050
AC36359-0250
TS36359-0010EA
140
USD
TS36359-0010
TS36359-0050
TS36359-0250
Hexachloroplatinic(IV) acid hydrate (38 - 40% Pt) 99.9% (trace metals basis)
Hexachloroplatinic(IV) acid hydrate
Formula:
H₂Cl₆Pt·H₂O MW: 427.83 g/mol Melting Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149909 CAS Number: 26023-84-7 UN: 2507 ADR: 8,III |
Specification Test Results
Appearance | Orange to orange-brown Powder and/or chunks |
Assay | 38 to 40 % (Pt) |
Total trace metal impurities | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...