Hexachloroplatinic(IV) acid hydrate ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate
AC40501-0010
AC40501-0050
AC40501-0250
TS40501-0010EA
142
USD
TS40501-0010
TS40501-0050
TS40501-0250
Hexachloroplatinic(IV) acid hydrate ACS
Hexachloroplatinic(IV) acid hydrate
Formula:
H₂Cl₆Pt·H₂O MW: 427.83 g/mol Melting Pt: 60 Storage Temperature: Ambient |
MDL Number:
MFCD00149909 CAS Number: 26023-84-7 EINECS: 241-010-7 UN: 2507 ADR: 8 III |
Specification Test Results
Appearance | Orange to orange-brown Powder and/or chunks |
Assay | ≥37.50 % (Pt) |
Solubility | (in alcohol) Passes test |
Suitability | (for K determination) Passes test |
Impurity | ≤0.05 % Alkali and other salts (as sulfates) |
Acidity (CH3COOH) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...