Hexachloroplatinic(IV) acid hydrate (∼40% Pt)
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19537-0010
AC19537-0050
AC19537-0250
TS19537-0010EA
122
USD
TS19537-0010
TS19537-0050
TS19537-0250
Hexachloroplatinic(IV) acid hydrate (∼40% Pt)
Hexachloroplatinic(IV) acid hydrate
Formula:
H₂Cl₆Pt·H₂O Melting Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149909 CAS Number: 26023-84-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III |
Specification Test Results
Appearance | Orange to orange-brown Powder and/or chunks |
Assay | 37.5 to 40.5 % (Pt) |
Total trace metal impurities | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...