Phosphotungstic acid hydrate

Supplier: Thermo Scientific Chemicals
Danger

AC20831-0025 AC20831-0250 AC20831-1000 AC20831-5000
TS20831-0025EA 2477 USD
TS20831-0025 TS20831-0250 TS20831-1000 TS20831-5000
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Formula: H₃[P(W₃O₁₀)₄]·nH₂O
Storage Temperature: Ambient
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,II
Merck Index: 13,07450

Order Now

Specification Test Results

Appearance White to yellow to beige to gray Powder or crystals
Assay ≥82 % (WO3)
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Ammonium (NH4) ≤50 ppm
Iron (Fe) ≤30 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤40 ppm
Loss on ignition ≤17 % (800°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR