Dodecamolybdophosphoric acid hydrate 80%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
AC20638-0250
AC20638-1000
AC20638-5000
TS20638-0250EA
75
USD
TS20638-0250
TS20638-1000
TS20638-5000
Dodecamolybdophosphoric acid hydrate 80%
Dodecamolybdophosphoric acid hydrate
Formula:
H₃[P(Mo₃O₁₀)₄]·nH₂O MW: 1843.27 g/mol Melting Pt: 78…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149913 CAS Number: 51429-74-4 EINECS: 234-713-5 UN: 3084 ADR: 8,II Merck Index: 12,07498 |
Specification Test Results
Appearance | Yellow to yellow-orange Adhering crystals |
Assay | ≥47.5 % (Mo) |
Loss on drying | ≤25 % |
Chloride (Cl) | ≤200 ppm |
Sulfate (SO4) | ≤250 ppm |
Ammonium (NH4) | ≤100 ppm |
Calcium (Ca) | ≤200 ppm |
Iron (Fe) | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...