Sodium diethyldithiocarbamate trihydrate ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

AC42431-1000 AC42431-5000
TS42431-1000EA 116 USD
TS42431-1000 TS42431-5000
Sodium diethyldithiocarbamate trihydrate ACS
Sodium diethyldithiocarbamate trihydrate
Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 95…98.5 °C
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Order Now

Specification Test Results

Appearance Off-white to light gray Powder or crystals
Infrared spectrum Conforms
Solubility (in water) Passes test
Sensitivity (to Copper) Passes test
Sodium (Na) 30.5 to 32.5 % (as Na2SO4)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR