Calcium acetate monohydrate ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Calcium di(acetate) monohydrate, Calcium acetate hydrate, Acetic acid calcium salt monohydrate
AC44635-1000
AC44635-5000
TS44635-1000EA
61
USD
TS44635-1000
TS44635-5000
Calcium acetate monohydrate ACS
Calcium acetate monohydrate
Formula:
(H₃CCOO)₂Ca·1H₂O Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Complexometric | ≥99.0 % |
Alkalinity | Passes test |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO4) | ≤0.01 % |
Barium (Ba) | ≤0.01 % |
Iron (Fe) | ≤0.001 % |
Magnesium (Mg) | ≤0.05 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.02 % |
Strontium (Sr) | ≤0.05 % |
Heavy metals (as Pb) | ≤0.005 % |
Titrable acid | ≤0.035 meq/g |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...