Hexamethyldisiloxane 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC19479-0025 AC19479-1000 AC19479-5000
TS19479-0025EA 265 USD
TS19479-0025 TS19479-1000 TS19479-5000
Hexamethyldisiloxane 98+%
Hexamethyldisiloxane
Formula: C₆H₁₈OSi₂
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: –59 °C
Density: 0.76 g/cm³
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008265
CAS Number: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3,II
REACH: 01-2119496108-31
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3765 to 1.3785 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR