1,1,3,3-Tetramethyldisiloxane 97%

Supplier: Thermo Scientific Chemicals
Danger

AC21651-0250 AC21651-1000 AC21651-5000
TS21651-0250EA 33 USD
TS21651-0250 TS21651-1000 TS21651-5000
1,1,3,3-Tetramethyldisiloxane 97%
1,1,3,3-Tetramethyldisiloxane
Formula: C₄H₁₄OSi₂
MW: 134.33 g/mol
Boiling Pt: 70…71 °C (760 mmHg)
Melting Pt: –78 °C
Density: 0.76 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008256
CAS Number: 3277-26-7
EINECS: 221-906-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3690 to 1.3710 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR