Platinium(IV) oxide 83% Pt
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19532-0010
AC19532-0050
TS19532-0010EA
208
USD
TS19532-0010
TS19532-0050
Platinium(IV) oxide 83% Pt
Platinium(IV) oxide
Formula:
O₂Pt Melting Pt: 450 °C |
MDL Number:
MFCD00011184 CAS Number: 1314-15-4 EINECS: 215-223-0 UN: 1479 ADR: 5.1,I Merck Index: 12,07686 |
Specification Test Results
Appearance | Brown Powder |
Assay | ≥79 % (Pt) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...