Cobalt(II) oxide , Co >74% -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt oxide

AC37920-0100 AC37920-1000
TS37920-0100EA 67 USD
TS37920-0100 TS37920-1000
Cobalt(II) oxide , Co >74% -325 mesh
Cobalt(II) oxide
Formula: CoO
MW: 74.93 g/mol
Melting Pt: 1933
Storage Temperature: Ambient
MDL Number: MFCD00016031
CAS Number: 1307-96-6
EINECS: 215-154-6
UN: 3288
ADR: 6.1 II
Merck Index: 12,02507

Order Now

Specification Test Results

Appearance Dark gray to green-brown Powder
Additional info oxides
Purity ≥74.0 % Co


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR