Rhodium(III) chloride Rh 48.6-49.2%, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Rhodium(III) chloride, Rhodium trichloride

AC41910-0025 AC41910-5000
TS41910-0025EA 2816 USD
TS41910-0025 TS41910-5000
Rhodium(III) chloride Rh 48.6-49.2%, anhydrous
Rhodium(III) chloride
Formula: Cl₃Rh
MW: 209.26 g/mol
Boiling Pt: 717 °C (730 mmHg)
Melting Pt: 450 °C
Density: 5.38 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011204
CAS Number: 10049-07-7
EINECS: 233-165-4
UN: 3260
ADR: 8,III
Merck Index: 12,08355

Order Now

Specification Test Results

Appearance Red, gray, red-black or black Crystalline powder, crystals or granules
Assay 48.6 to 49.2 % (Rh)
Total trace metal impurities ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR