Palladium(II) chloride 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium dichloride

AC36967-0250 369670010 369670050
TS36967-0010EA 168.23 USD
TS36967-0010 TS36967-0050 TS36967-0250 TS369670010 TS369670050
Palladium(II) chloride 99.999% (trace metals basis)
Palladium(II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678…680 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Specification Test Results

Appearance Brown Powder
Assay 59 to 61 % (Pd)
Total trace metal impurities ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR