Palladium(II) chloride (59% Pd)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium dichloride

AC19520-0010 AC19520-0050 AC19520-0250
TS19520-0010EA 199 USD
TS19520-0010 TS19520-0050 TS19520-0250
Palladium(II) chloride (59% Pd)
Palladium(II) chloride
Formula: PdCl₂
Melting Pt: 678…680 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Specification Test Results

Appearance Red-brown Powder
Assay 59 to 60 % (Pd)
Calcium (Ca) ≤30 ppm
Copper (Cu) ≤15 ppm
Iron (Fe) ≤15 ppm
Lead (Pb) ≤15 ppm
Magnesium (Mg) ≤15 ppm
Silicon (Si) ≤30 ppm
Silver (Ag) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR