Tungsten trioxide 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tungsten (VI) oxide

AC21421-1000 AC21421-5000
TS21421-1000EA 85 USD
TS21421-1000 TS21421-5000
Tungsten trioxide 99+%
Tungsten trioxide
Formula: O₃W
MW: 231.84
Melting Pt: 1473
Density: 7.16
Storage Temperature: Ambient
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4
Merck Index: 12,09947

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Assay ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR