Tungsten(VI) chloride 99.9+% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC22143-0100 AC22143-1000
TS22143-0100EA 61 USD
TS22143-0100 TS22143-1000
Tungsten(VI) chloride 99.9+% (trace metals basis)
Tungsten(VI) chloride
Formula: Cl₆W
Boiling Pt: 347 °C (760 mmHg)
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Gray to dark blue to violet to black Powder
Total trace metal impurities ≤0.10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR