Tungsten(VI) chloride 99% 20 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tungsten hexachloride

AC20030-0100 AC20030-0500 AC20030-2500
TS20030-0100EA 47 USD
TS20030-0100 TS20030-0500 TS20030-2500
Tungsten(VI) chloride 99% 20 mesh
Tungsten(VI) chloride
Formula: Cl₆W
MW: 396.56 g/mol
Boiling Pt: 347
Melting Pt: 275
Storage Temperature: Ambient
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8 II

Order Now

Specification Test Results

Appearance Gray to dark blue to violet to black Powder
Purity ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR