Chromium(II) chloride 90%

Supplier: Thermo Scientific Chemicals
Warning

AC22381-0050 AC22381-0100
TS22381-0050EA 121 USD
TS22381-0050 TS22381-0100
Chromium(II) chloride 90%
Chromium(II) chloride
Formula: Cl₂Cr
Boiling Pt: 1302 °C (1013 hPa)
Melting Pt: 824 °C
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
Merck Index: 13,02264

Order Now

Specification Test Results

Appearance Green to gray Powder
Titration Argentometric ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR