Tantalum pentachloride 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC20391-0050 AC20391-0250 AC20391-1000
TS20391-0050EA 48 USD
TS20391-0050 TS20391-0250 TS20391-1000
Tantalum pentachloride 99.99% (trace metals basis)
Tantalum pentachloride
Formula: Cl₅Ta
Boiling Pt: 232…235 °C (1013 hPa)
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Total trace metal impurities ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR