Diyttrium trioxide 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

AC19461-0100 AC19461-0500 AC19461-2500
TS19461-0100EA 24 USD
TS19461-0100 TS19461-0500 TS19461-2500
Diyttrium trioxide 99.99% (trace metals basis)
Diyttrium trioxide
Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Specification Test Results

Appearance White Powder
Assay ≥99.99 % (Y2O3) TREO base
Particle size 2 to 10 µm (d50)
Total conc. of imp.as REO ≤100 ppm
Loss on ignition ≤1.0 % (1000°C) Material base


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR