Silver nitrite 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21069-0100
AC21069-0500
TS21069-0100EA
74
USD
TS21069-0100
TS21069-0500
Silver nitrite 99%
Silver nitrite
Formula:
AgNO₂ Melting Pt: 140 °C Density: 4.45 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003413 CAS Number: 7783-99-5 EINECS: 232-041-7 UN: 2627 ADR: 5.1,II Merck Index: 12,08663 |
Specification Test Results
Appearance | Yellow to yellow-green Powder or crystals and/or chunks |
Titration Argentometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...