Gold trichloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gold(III) chloride, Gold (III) chloride

AC31864-0050 AC31864-5000
TS31864-0050EA 913 USD
TS31864-0050 TS31864-5000
Gold trichloride 99%
Gold trichloride
Formula: AuCl₃
MW: 303.32
Melting Pt: 254 °C
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00014172
CAS Number: 13453-07-1
EINECS: 236-623-1
Merck Index: 13,04536

Order Now

Specification Test Results

Appearance Orange-red to brown to violet Crystalline powder
Assay ≥98.5 % (ex Au)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR