Ammonium cerium (IV) sulfate dihydrate 95% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetraammonium cerium tetrakis(sulphate) dihydrate, Cerium (IV) ammonium sulphate dihydrate, Tetraammonium cerium(IV) sulphate dihydrate, Ammonium cerium(IV) sulphate hydrate, Ammonium ceric sulphate dihydrate

AC32579-0025 AC32579-1000 AC32579-5000
TS32579-0025EA 1489 USD
TS32579-0025 TS32579-1000 TS32579-5000
Ammonium cerium (IV) sulfate dihydrate 95% ACS
Ammonium cerium(IV) sulphate dihydrate
Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
MW: 632.55 g/mol
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00149417
CAS Number: 10378-47-9
EINECS: 231-567-4

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Titration ≥94 %
Phosphate (PO4) ≤0.03 %
Iron (Fe) ≤0.01 %
Insoluble matter ≤0.05 % (in dilute sulfuric acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR