Europium(III) oxide 99.99% (trace metals basis)
Supplier: Thermo Scientific Chemicals
AC19302-0010
AC19302-0050
AC19302-0250
TS19302-0010EA
49
USD
TS19302-0010
TS19302-0050
TS19302-0250
Europium(III) oxide 99.99% (trace metals basis)
Europium(III) oxide
Formula:
Eu₂O₃ MW: 351.92 g/mol Density: 7.42 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010997 CAS Number: 1308-96-9 EINECS: 215-165-6 Merck Index: 14,03904 |
Specification Test Results
Appearance | White to light pink Powder |
Total conc. of imp.as REO | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...