Terbium(III) chloride hexahydrate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals

AC19961-0050 AC19961-0250
TS19961-0050EA 91 USD
TS19961-0050 TS19961-0250
Terbium(III) chloride hexahydrate 99.9% (trace metals basis)
Terbium(III) chloride hexahydrate
Formula: Cl₃Tb·6H₂O
MW: 373.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149870
CAS Number: 13798-24-8
EINECS: 233-132-4
Merck Index: 12,09300

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Total conc. of imp.as REO ≤0.1 %
Terbium (Tb) 42.13 to 42.99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR