Tetrabutylammonium fluoride 75% in water

Supplier: Thermo Scientific Chemicals
Danger

AC45180-0250 AC45180-1000 AC45180-5000
TS45180-0250EA 66 USD
TS45180-0250 TS45180-1000 TS45180-5000
Tetrabutylammonium fluoride 75% in water
Tetrabutylammonium fluoride
Formula: C₁₆H₃₆FN
MW: 261.47
Density: 0.953 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011747
CAS Number: 429-41-4
UN: 2922
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless to light yellow Viscous liquid
Water 20 to 30 %
Titration with HClO4 70 to 80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR