Aluminium ammonium sulphate dodecahydrate 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aluminium ammonium bis(sulphate) dodecahydrate, Ammonium alum, Ammonium aluminum sulphate dodecahydrate, Aluminum ammonium sulphatedodecahydrate

AC20626-0010 AC20626-0025 AC20626-1000
TS20626-0010EA 259 USD
TS20626-0010 TS20626-0025 TS20626-1000
Aluminium ammonium sulphate dodecahydrate 99%, pure
Aluminium ammonium sulphate dodecahydrate
Formula: AlNH₄(SO₄)₂·12H₂O
MW: 453.33 g/mol
Boiling Pt: 200
Melting Pt: 93
Density: 1.64 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00051042
CAS Number: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 98.5 to 102.5 %
Loss on drying 45 % to 48 % (2 g, 300°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR