Diethylaluminium chloride 1 M in hexanes, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC19952-1000 AC19952-8000
TS19952-1000EA 64 USD
TS19952-1000 TS19952-8000
Diethylaluminium chloride 1 M in hexanes, AcroSeal™
Diethylaluminium chloride
Formula: C₄H₁₀AlCl
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00000459
CAS Number: 96-10-6
EINECS: 202-477-2
UN: 3394
ADR: 4.2,I

Order Now

Specification Test Results

Appearance Colorless Solution
Strength 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR