Aluminium sulfate 99.999% (trace metals basis), Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC19243-0050 AC19243-0250 AC19243-1000
TS19243-0050EA 58 USD
TS19243-0050 TS19243-0250 TS19243-1000
Aluminium sulfate 99.999% (trace metals basis), Extra Pure
Aluminium sulphate
Formula: Al₂O₁₂S₃
Melting Pt: 770 °C
Storage Temperature: Ambient
MDL Number: MFCD00003423
CAS Number: 10043-01-3
EINECS: 233-135-0
Merck Index: 12,00381

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Titration Complexometric ≥15 % (Al) Typical
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR