Aluminium hydroxide, powder, Extra Pure
Supplier: Thermo Scientific Chemicals
AC21913-0020
AC21913-0100
AC21913-0250
AC21913-5000
TS21913-0020EA
160
USD
TS21913-0020
TS21913-0100
TS21913-0250
TS21913-5000
Aluminium hydroxide, powder, Extra Pure
Aluminium hydroxide
Formula:
Al(OH)₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003420 CAS Number: 21645-51-2 EINECS: 244-492-7 Merck Index: 12,00355 |
Specification Test Results
Appearance | White to almost white Powder |
Assay | ≥63.5 % (Al2O3) |
Soluble matter | ≤0.2 % (in water) |
Loss on ignition | ≤36.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...