Magnesium hydroxide carbonate light (∼40.0 - 45.0% MgO) for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic

AC21107-0010 AC21107-0025
TS21107-0010EA 38 USD
TS21107-0010 TS21107-0025
Magnesium hydroxide carbonate light (∼40.0 - 45.0% MgO) for analysis
Magnesium hydroxide carbonate
Formula: MgCO₃ (linear)
MW: 84.31 g/mol
Density: 2.16 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149783
CAS Number: 12125-28-9
EINECS: 235-192-7

Order Now

Specification Test Results

Appearance White Powder
Assay 40.0 to 45.0 % (MgO)
Chloride (Cl) ≤700 ppm
Sulfate (SO4) ≤3000 ppm
Arsenic (As) ≤2 ppm
Calcium (Ca) ≤7500 ppm
Iron (Fe) ≤400 ppm
Heavy metals (as Pb) ≤20 ppm
Soluble matter ≤1 % (in water)
Insoluble matter ≤0.05 % (in acetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR