Magnesium perchlorate ACS desiccant

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Anhydrone

AC41346-1000 AC41346-5000
TS41346-1000EA 88 USD
TS41346-1000 TS41346-5000
Magnesium perchlorate ACS desiccant
Magnesium perchlorate
Formula: Cl₂MgO₈
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00011102
CAS Number: 10034-81-8
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II
Merck Index: 13,05702

Order Now

Specification Test Results

Appearance White Crystalline powder or flakes and/or chunks
Loss on drying ≤8 % (1 g, 190°C)
Suitability (for moisture absorption) Passes test
Titrable base ≤0.025 meq/g
Titrable free acid ≤0.005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR