Decamethylcyclopentasiloxane 97%

Supplier: Thermo Scientific Chemicals

AC45458-0500 AC45458-2500
TS45458-0500EA 35 USD
TS45458-0500 TS45458-2500
Decamethylcyclopentasiloxane 97%
Formula: C₁₀H₃₀O₅Si₅
Boiling Pt: 101 °C (20 mmHg)
Density: 0.959 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00046966
CAS Number: 541-02-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR