Sodium methanesulfonate 99%

Supplier: Thermo Scientific Chemicals

AC44211-0050 AC44211-1000 AC44211-5000
TS44211-0050EA 46 USD
TS44211-0050 TS44211-1000 TS44211-5000
Sodium methanesulfonate 99%
Sodium methanesulphonate
Formula: CH₃NaO₃S
MW: 118.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064389
CAS Number: 2386-57-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Titration after Ion exchange ≥98.5 %
Free acid ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR