Aluminium isopropoxide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

AC20517-0010 AC20517-0051 AC20517-5000
TS20517-0010EA 75 USD
TS20517-0010 TS20517-0051 TS20517-5000
Aluminium isopropoxide 98%
Aluminium isopropoxide
Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Storage Temperature: Ambient
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 13,00346

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 129 to 136 °C
Aluminium (Al) 12.9 to 13.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR