Sodium trifluoromethanesulphonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Trifluoromethanesulphonic acid sodium salt
AC35875-0050
AC35875-0250
TS35875-0050EA
50
USD
TS35875-0050
TS35875-0250
Sodium trifluoromethanesulphonate 98%
Sodium trifluoromethanesulphonate
Formula:
CF₃NaO₃S MW: 172.06 g/mol Melting Pt: 253…255 °C |
MDL Number:
MFCD00061607 CAS Number: 2926-30-9 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Assay | ≥97.5 % (ex Na) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...