Triisopropylsilyl trifluoromethanesulphonate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate

AC25070-0100 AC25070-0500
TS25070-0100EA 83 USD
TS25070-0100 TS25070-0500
Triisopropylsilyl trifluoromethanesulphonate 97%
Triisopropylsilyl trifluoromethanesulphonate
Formula: C₁₀H₂₁F₃O₃SSi
MW: 306.42 g/mol
Boiling Pt: 120
Density: 1.14 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4160 (20°C, 589 nm)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR