Silver(I) acetate 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silver acetate, Acetic acid silver (I) salt
AC20239-0250
AC20239-1000
AC20239-5000
TS20239-0250EA
188
USD
TS20239-0250
TS20239-1000
TS20239-5000
Silver(I) acetate 99%, pure
Silver(I) acetate
Formula:
C₂H₃AgO₂ MW: 166.91 g/mol Melting Pt: 2000 Density: 3.25 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012446 CAS Number: 563-63-3 EINECS: 209-254-9 UN: 3077 ADR: 9 III Merck Index: 12,08648 |
Specification Test Results
Appearance | Off-white to beige to light gray Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...