Platinium(IV) oxide monohydrate 99.8% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC21193-0010 AC21193-0050 AC21193-2500
TS21193-0010EA 278 USD
TS21193-0010 TS21193-0050 TS21193-2500
Platinium(IV) oxide monohydrate 99.8% (trace metals basis)
Platinium(IV) oxide monohydrate
Formula: O₂Pt·1H₂O
MW: 245.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066964
CAS Number: 12137-21-2
EINECS: 215-223-0
UN: 1479
ADR: 5.1,I

Order Now

Specification Test Results

Appearance Brown to black Powder
Surface area ≥80 m²/g
Total trace metal impurities ≤2000 ppm
Platinum (Pt) 77.6 to 82 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR