Ruthenium(III) chloride hydrate (35 - 40% Ru)

Supplier: Thermo Scientific Chemicals
Danger

AC19548-0010 AC19548-0050 AC19548-0250 AC19548-1000
TS19548-0010EA 60 USD
TS19548-0010 TS19548-0050 TS19548-0250 TS19548-1000
Ruthenium(III) chloride hydrate (35 - 40% Ru)
Ruthenium(III) chloride hydrate
Formula: Cl₃Ru·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance Dark brown to black Powder
Assay 35 to 40 % (Ru)
Additional info Trace analysis values are based on Ru
Calcium (Ca) ≤300 ppm
Copper (Cu) ≤300 ppm
Gold (Au) ≤300 ppm
Iridium (Ir) ≤500 ppm
Iron (Fe) ≤300 ppm
Magnesium (Mg) ≤300 ppm
Osmium (Os) ≤100 ppm
Palladium (Pd) ≤500 ppm
Platinum (Pt) ≤500 ppm
Potassium (K) ≤300 ppm
Rhodium (Rh) ≤500 ppm
Silicon (Si) ≤300 ppm
Silver (Ag) ≤300 ppm
Sodium (Na) ≤300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR