Rhodium(III) chloride hydrate 38% Rh

Supplier: Thermo Scientific Chemicals
Danger

AC19552-0010 AC19552-0050
TS19552-0010EA 990 USD
TS19552-0010 TS19552-0050
Rhodium(III) chloride hydrate 38% Rh
Rhodium(III) chloride hydrate
Formula: Cl₃Rh·H₂O
Boiling Pt: 800 °C (1013 hPa)
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149839
CAS Number: 20765-98-4
EINECS: 233-165-4
UN: 3260

Order Now

Specification Test Results

Appearance Red-brown to dark brown Crystalline powder
Assay 37 to 40 % (Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR