Iridium(III) chloride trihydrate (53 - 56% Ir)

Supplier: Thermo Scientific Chemicals

AC19550-0010 AC19550-0050 AC19550-0250
TS19550-0010EA 448 USD
TS19550-0010 TS19550-0050 TS19550-0250
Iridium(III) chloride trihydrate (53 - 56% Ir)
Iridium(III) chloride trihydrate
Formula: Cl₃Ir·₃H₂O
MW: 352.62
Melting Pt: 763 (dec.)
Density: 5.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149750
CAS Number: 13569-57-8
EINECS: 233-044-6

Order Now

Specification Test Results

Appearance Green to black Crystals
Assay 53 to 56 % (Ir)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR