Yttrium(III) chloride hexahydrate 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC19918-0500
AC19918-2500
TS19918-0500EA
87
USD
TS19918-0500
TS19918-2500
Yttrium(III) chloride hexahydrate 99.9% (trace metals basis)
Yttrium(III) chloride hexahydrate
Formula:
YCl₃·6H₂O Boiling Pt: 1507 °C (1013 hPa) Melting Pt: 100 °C |
MDL Number:
MFCD00149941 CAS Number: 10025-94-2 EINECS: 233-801-0 |
Specification Test Results
Appearance | White Adhering crystalline powder or crystals |
Total trace metal impurities | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...