Mercury(II) nitrate monohydrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury(II) nitrate hydrate, Mercury dinitrate monohydrate

AC21314-0010 AC21314-0500 AC21314-2500
TS21314-0010EA 899 USD
TS21314-0010 TS21314-0500 TS21314-2500
Mercury(II) nitrate monohydrate 98+%
Mercury(II) nitrate monohydrate
Formula: Hg(NO₃)₂·1H₂O
MW: 342.62 g/mol
Melting Pt: 79
Density: 4.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149736
CAS Number: 7783-34-8
EINECS: 233-152-3
UN: 1625
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White Adhering crystalline powder
Titration Complexometric ≥98 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤20 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤20 ppm
Mercurous Mercury (as Hg) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR