Ytterbium(III) chloride hexahydrate 99.99+% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

AC43718-0100 AC43718-0500
TS43718-0100EA 123 USD
TS43718-0100 TS43718-0500
Ytterbium(III) chloride hexahydrate 99.99+% (trace metals basis)
Ytterbium(III) chloride hexahydrate
Formula: Cl₃Yb·6H₂O
MW: 387.49 g/mol
Melting Pt: 865 °C
Storage Temperature: Ambient
MDL Number: MFCD00149885
CAS Number: 10035-01-5
EINECS: 233-800-5

Order Now

Specification Test Results

Appearance White Powder or crystals
Total trace metal impurities ≤100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR