Ytterbium(III) chloride hexahydrate 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC19443-0010
AC19443-0100
AC19443-0500
TS19443-0010EA
36
USD
TS19443-0010
TS19443-0100
TS19443-0500
Ytterbium(III) chloride hexahydrate 99.9% (trace metals basis)
Ytterbium(III) chloride hexahydrate
Formula:
Cl₃Yb·6H₂O MW: 387.49 g/mol Melting Pt: 865 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149885 CAS Number: 10035-01-5 EINECS: 233-800-5 |
Specification Test Results
Appearance | White Crystalline powder or crystals and/or chunks |
Total conc. of imp.as REO | ≤0.1 % |
Ytterbium (Yb) | 43.1 to 46.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...